Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp638 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166371.1 | NP_001159843.1 | 2 | 862 | ||
NM_008717.3 | NP_032743.2 | 2 | 872 | |||
GenBank mRNA | AK043029.1 | - | 2 | 844 | ||
AK157791.1 | - | 2 | 853 | |||
AK163476.1 | - | 1 | 468 | |||
AK163955.1 | - | 2 | 824 | |||
AK165189.1 | - | 2 | 843 | |||
BC019223.1 | - | 2 | 844 | |||
BC076615.1 | - | 2 | 825 | 30640388 | ||
D83033.1 | - | 2 | 858 |