Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nppc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010933.5 | NP_035063.1 | 3 | 882 | ||
GenBank mRNA | AK020093.1 | - | 3 | 888 | ||
AK036176.1 | - | 3 | 886 |