Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008727.5 | NP_032753.5 | 5, 6 | 1621 | ||
GenBank mRNA | BC110659.1 | - | 5, 6 | 1501 | 30847317 | |
BC139767.1 | - | 5, 6 | 1480 | |||
J05504.1 | - | 6, 7 | 1605 | |||
L31932.1 | - | 5, 6 | 1269 |