Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nsd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008739.3 | NP_032765.3 | 12 | 4858 | ||
GenBank mRNA | BQ442794.1 | - | 5 | 529 | ||
CB247038.1 | - | 4 | 409 |