Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Musk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037127.2 | NP_001032204.2 | 8 | 1033 | ||
NM_001037128.1 | NP_001032205.1 | 8 | 1033 | |||
NM_001037129.1 | NP_001032206.1 | 8 | 1033 | |||
NM_001037130.1 | NP_001032207.1 | 7 | 1003 | |||
NM_001165996.1 | NP_001159468.1 | 7 | 1003 | |||
NM_010944.2 | NP_035074.2 | 7 | 1003 | |||
GenBank mRNA | AB691772.1 | - | 7 | 853 | ||
AY360453.1 | - | 7 | 898 | |||
AY955296.1 | - | 8 | 883 | |||
AY955297.1 | - | 8 | 883 | |||
AY955298.1 | - | 8 | 883 | |||
AY956403.1 | - | 7 | 853 | |||
AY956404.1 | - | 7 | 853 | |||
BC100342.1 | - | 8 | 957 | 30935290 | ||
U37708.1 | - | 7 | 1001 | |||
U37709.1 | - | 7 | 1001 | |||
X86444.1 | - | 7 | 898 |