Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nsmaf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010945.2 | NP_035075.2 | 31 | 2917 | ||
GenBank mRNA | AF013632.1 | - | 31 | 2910 | ||
AK172560.1 | - | 5 | 499 | |||
BC066075.1 | - | 30 | 3812 | |||
BC082810.1 | - | 30 | 3812 | |||
BC145961.1 | - | 31 | 2731 | 40131264 | ||
BC145963.1 | - | 31 | 2731 | 40131266 |