Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nsmaf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010945.2 | NP_035075.2 | 20, 21 | 1848 | ||
GenBank mRNA | AF013632.1 | - | 20, 21 | 1841 | ||
AK040369.1 | - | 4, 5 | 4158 | |||
AK049887.1 | - | 20, 21 | 1800 | |||
AK147709.1 | - | 20, 21 | 1709 | |||
AK164305.1 | - | 20, 21 | 1850 | |||
BC066075.1 | - | 19, 20 | 2743 | |||
BC082810.1 | - | 19, 20 | 2743 | |||
BC145961.1 | - | 20, 21 | 1662 | 40131264 | ||
BC145963.1 | - | 20, 21 | 1662 | 40131266 |