Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ntan1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010946.3 | NP_035076.1 | 10 | 896 | ||
GenBank mRNA | AA161752.1 | - | 1 | 16 | ||
AK206842.1 | - | 1 | 54 | |||
AK208313.1 | - | 1 | 54 | |||
BC030172.1 | - | 10 | 894 | 5037501 | ||
U57692.1 | - | 10 | 850 |