Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nthl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB006812.1 | - | 5 | 775 | ||
BC138852.1 | - | 5 | 792 | 8861874 | ||
BC138855.1 | - | 5 | 792 | 8861877 | ||
BC145443.1 | - | 5 | 701 | |||
Y09688.1 | - | 5 | 801 |