Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ntrk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025074.2 | NP_001020245.1 | 4, 5 | 1023 | ||
NM_001282961.1 | NP_001269890.1 | 4, 5 | 990 | |||
NM_008745.3 | NP_032771.1 | 4, 5 | 990 | |||
GenBank mRNA | AB377224.1 | - | 3, 4 | 400 | ||
AB377225.1 | - | 3, 4 | 400 | |||
AK009606.1 | - | 3, 4 | 393 | |||
AK018789.1 | - | 4, 5 | 921 | |||
AK028827.1 | - | 4, 5 | 843 | |||
AK147391.1 | - | 4, 5 | 821 | |||
AK147441.1 | - | 3, 4 | 811 | |||
AK160789.1 | - | 4, 5 | 991 | |||
BC052014.1 | - | 3, 4 | 785 | 5707891 | ||
KF982303.1 | - | 2, 3 | 129 | |||
M33385.1 | - | 3, 4 | 362 | |||
X17647.1 | - | 873 |