Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ntrk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008746.5 | NP_032772.3 | 6 | 781 | ||
NM_182809.2 | NP_877961.1 | 6 | 781 | |||
GenBank mRNA | AF035399.1 | - | 7 | 876 | ||
AF035400.1 | - | 6 | 930 | |||
AY336094.1 | - | 6 | 726 | |||
BC139764.1 | - | 7 | 793 | 40110345 |