Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nucb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163662.1 | NP_001157134.1 | 5 | 561 | ||
NM_008749.2 | NP_032775.1 | 5 | 561 | |||
GenBank mRNA | AK002630.1 | - | 5 | 542 | ||
AK004886.1 | - | 5 | 532 | |||
AK007976.1 | - | 5 | 535 | |||
AK045087.1 | - | 5 | 561 | |||
AK140710.1 | - | 5 | 539 | |||
AK160981.1 | - | 5 | 548 | |||
AK164088.1 | - | 5 | 543 | |||
AK170717.1 | - | 5 | 556 | |||
AK182386.1 | - | 1 | 26 | |||
BC072554.1 | - | 5 | 539 | 5684979 | ||
M96823.1 | - | 4 | 483 |