Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Numb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136075.2 | NP_001129547.1 | 4, 5 | 328 | ||
NM_001272055.1 | NP_001258984.1 | 3, 4 | 295 | |||
NM_001272056.1 | NP_001258985.1 | 5, 6 | 525 | |||
NM_010949.2 | NP_035079.1 | 4, 5 | 328 | |||
GenBank mRNA | AF169191.1 | - | 3, 4 | 280 | ||
AF169192.1 | - | 4, 5 | 313 | |||
AF170709.1 | - | 4, 5 | 313 | |||
AK004553.1 | - | 6, 7 | 550 | |||
BC033459.1 | - | 6, 7 | 566 | 3991630 | ||
BC050108.1 | - | 7, 8 | 585 | |||
U70674.1 | - | 5, 6 | 446 |