Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Numbl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010950.2 | NP_035080.2 | 3 | 307 | ||
GenBank mRNA | AK147523.1 | - | 1 | 6094 | ||
BC059915.1 | - | 2 | 896 | |||
BC068116.1 | - | 3 | 134 | 6809443 | ||
BC098097.1 | - | 3 | 148 | 6403012 | ||
CJ070714.1 | - | 3 | 307 | |||
U96441.1 | - | 3 | 191 |