Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PI4KA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_058004.3 | NP_477352.3 | 43 | 5171 | ||
XM_005261634.1 | XP_005261691.1 | 42 | 5078 | |||
XM_017028829.1 | XP_016884318.1 | 42 | 5105 | |||
XM_017028830.1 | XP_016884319.1 | 29 | 3327 | |||
GenBank mRNA | AB210002.1 | - | 43 | 5141 | ||
AF012872.1 | - | 42 | 4910 | |||
AK095811.1 | - | 3 | 261 | |||
AK124692.1 | - | 1 | 680 | |||
AK302424.1 | - | 43 | 5099 | |||
AK314781.1 | - | 15 | 1825 | |||
BC018120.1 | - | 13 | 1488 | |||
BC053654.1 | - | 12 | 1464 | |||
L36151.1 | - | 11 | 1457 |