Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016978.2 | NP_058674.1 | 2 | 271 | ||
GenBank mRNA | AK128914.1 | - | 2 | 262 | ||
AK145877.1 | - | 2 | 272 | |||
AK146413.1 | - | 2 | 216 | |||
AK168851.1 | - | 2 | 220 | |||
AK185243.1 | - | 1 | 55 | |||
AK185430.1 | - | 1 | 55 | |||
BC008119.1 | - | 2 | 204 | 3498240 | ||
X64837.1 | - | 1 | 245 |