Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016978.2 | NP_058674.1 | 9 | 1177 | ||
GenBank mRNA | AK128914.1 | - | 9 | 1168 | ||
AK145877.1 | - | 9 | 1178 | |||
AK146413.1 | - | 9 | 1122 | |||
AK168851.1 | - | 9 | 1126 | |||
AK194444.1 | - | 34 | ||||
BC008119.1 | - | 9 | 1110 | 3498240 | ||
X64837.1 | - | 8 | 1151 |