Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Obp1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008754.2 | NP_032780.2 | 4 | 358 | ||
GenBank mRNA | AK017455.1 | - | 4 | 359 | ||
BC125514.1 | - | 4 | 345 | 40130029 | ||
BC125516.1 | - | 4 | 345 | 40130031 |