Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ocm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033039.3 | NP_149028.2 | 4 | 405 | ||
GenBank mRNA | BQ567296.1 | - | 4 | 295 |