Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Odc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013614.2 | NP_038642.2 | 4 | 674 | ||
GenBank mRNA | AK077679.1 | - | 4 | 576 | ||
AK139610.1 | - | 4 | 576 | |||
AK150283.1 | - | 4 | 577 | |||
AK150343.1 | - | 4 | 576 | |||
AK152525.1 | - | 4 | 576 | |||
AK168219.1 | - | 4 | 576 | |||
AK169214.1 | - | 4 | 576 | |||
AK172568.1 | - | 4 | 576 | |||
BC033264.1 | - | 4 | 565 | 2645291 | ||
BC059826.1 | - | 4 | 555 | 6808137 | ||
BC083122.1 | - | 4 | 562 | 6329274 | ||
EU684749.1 | - | 2 | 264 | |||
M20617.2 | - | 2 | 280 | |||
S64539.1 | - | 4 | 574 |