Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Odc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013614.2 | NP_038642.2 | 10 | 1379 | ||
GenBank mRNA | AK077679.1 | - | 10 | 1282 | ||
AK139610.1 | - | 10 | 1281 | |||
AK150283.1 | - | 10 | 1281 | |||
AK150343.1 | - | 10 | 1280 | |||
AK152525.1 | - | 10 | 1279 | |||
AK166093.1 | - | 10 | 1286 | |||
AK168219.1 | - | 10 | 1280 | |||
AK169214.1 | - | 10 | 1280 | |||
AK172568.1 | - | 10 | 1282 | |||
AK190288.1 | - | 1 | 67 | |||
AK194223.1 | - | 1 | 82 | |||
AK194907.1 | - | 1 | 74 | |||
AK201555.1 | - | 1 | 78 | |||
AK202063.1 | - | 67 | ||||
AK202843.1 | - | 1 | 67 | |||
BC033264.1 | - | 10 | 1270 | 2645291 | ||
BC059826.1 | - | 10 | 1260 | 6808137 | ||
BC083122.1 | - | 10 | 1267 | 6329274 | ||
EU684749.1 | - | 8 | 969 | |||
M12330.1 | - | 5 | 438 | |||
M12331.1 | - | 1 | 14 | |||
M20617.2 | - | 8 | 985 | |||
S64539.1 | - | 10 | 1279 |