Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ICAM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320605.1 | NP_001307534.1 | 4 | 1087 | ||
NM_001320606.1 | NP_001307535.1 | 5 | 1672 | |||
NM_001320608.1 | NP_001307537.1 | 4 | 1692 | |||
NM_002162.4 | NP_002153.2 | 5 | 1375 | |||
GenBank mRNA | AF340060.1 | - | 5 | 1128 | ||
AF340061.1 | - | 5 | 1128 | |||
AK226023.1 | - | 5 | 1209 | |||
AK309643.1 | - | 5 | 1457 | |||
AK312826.1 | - | 5 | 1210 | |||
AX746321.1 | - | 5 | 1170 | |||
BC040123.1 | - | 5 | 1155 | |||
BC046121.1 | - | 3 | 1802 | |||
BC058903.1 | - | 5 | 1161 | 5207143 | ||
S50015.1 | - | 5 | 1156 | |||
X69711.1 | - | 5 | 1170 | |||
X69819.1 | - | 5 | 1163 |