Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ogn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008760.4 | NP_032786.1 | 6 | 819 | ||
GenBank mRNA | AK053650.1 | - | 6 | 820 | ||
AK075638.1 | - | 5 | 777 | |||
AK160760.1 | - | 6 | 841 | |||
AK163772.1 | - | 5 | 771 | |||
BC021939.1 | - | 5 | 722 | 5067073 | ||
D31951.1 | - | 5 | 732 |