Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ogn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008760.4 | NP_032786.1 | 5 | 694 | ||
GenBank mRNA | AK028300.1 | - | 5 | 720 | ||
AK053650.1 | - | 5 | 695 | |||
AK075638.1 | - | 4 | 651 | |||
AK160760.1 | - | 5 | 716 | |||
AK163772.1 | - | 4 | 646 | |||
BC021939.1 | - | 4 | 597 | 5067073 | ||
D31951.1 | - | 4 | 607 |