Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146783.2 | NP_666994.2 | 1 | 510 | ||
GenBank mRNA | BC130238.1 | - | 1 | 510 | 40142117 | |
X89681.1 | - | 1 | 132 |