Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147027.2 | NP_667238.2 | 1 | 422 | ||
GenBank mRNA | AF102527.1 | - | 1 | 236 | ||
BC106818.1 | - | 1 | 418 | |||
BC106819.1 | - | 1 | 422 | 40043407 | ||
BC106824.1 | - | 1 | 422 | 40043472 | ||
X89691.1 | - | 1 | 51 |