Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303265.1 | NP_001290194.1 | 2, 3 | 251 | ||
NM_001545.2 | NP_001536.1 | 2, 3 | 251 | |||
XM_017024574.1 | XP_016880063.1 | 2, 3 | 244 | |||
GenBank mRNA | AK222553.1 | - | 2, 3 | 241 | ||
AK314138.1 | - | 2, 3 | 239 | |||
BC015335.1 | - | 2, 3 | 234 | 4418983 | ||
BI870683.1 | - | 2, 3 | 108 | |||
BT007111.1 | - | 2, 3 | 228 | |||
DB456649.1 | - | 2, 3 | 251 | |||
X81788.1 | - | 2, 3 | 230 |