Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146370.1 | NP_666482.1 | 1 | 104 | ||
GenBank mRNA | BC120658.1 | - | 1 | 136 | 40129581 | |
BC120660.1 | - | 1 | 136 | 40129583 |
Gene Symbol: Olfr434 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146369.1 | NP_666481.1 | 1 | 104 | ||
GenBank mRNA | BC120611.1 | - | 1 | 136 | 40129534 | |
BC120613.1 | - | 1 | 136 | 40129536 |