Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010991.2 | NP_035121.1 | 1 | 934 | ||
GenBank mRNA | BC145891.1 | - | 1 | 934 | 40131194 | |
BC145893.1 | - | 1 | 934 | 40131196 |