Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010997.1 | NP_035127.1 | 1 | 402 | ||
GenBank mRNA | AF102530.1 | - | 1 | 207 | ||
BC099884.1 | - | 1 | 401 | |||
BC099885.1 | - | 1 | 401 | |||
BC100678.1 | - | 1 | 401 |