Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ID2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002166.4 | NP_002157.2 | 2 | 546 | ||
GenBank mRNA | AK222682.1 | - | 2 | 473 | ||
AK311988.1 | - | 2 | 473 | |||
AY634687.1 | - | 2 | 482 | |||
BC030639.2 | - | 2 | 473 | 4820416 | ||
D13891.1 | - | 2 | 459 | |||
M97796.1 | - | 2 | 471 |