Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146861.1 | NP_667072.1 | 1 | 602 | ||
GenBank mRNA | BC116994.1 | - | 1 | 667 | 40126313 | |
BC119365.1 | - | 1 | 667 | 8734114 |
Gene Symbol: Olfr763 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146862.1 | NP_667073.1 | 1 | 602 | ||
GenBank mRNA | BC119229.1 | - | 1 | 667 | 8733978 | |
BC119231.1 | - | 1 | 667 | 8733980 |