Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PI4KA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_058004.3 | NP_477352.3 | 55 | 6656 | ||
XM_005261634.1 | XP_005261691.1 | 54 | 6563 | |||
XM_005261635.1 | XP_005261692.1 | 48 | 5828 | |||
XM_017028829.1 | XP_016884318.1 | 54 | 6590 | |||
XM_017028830.1 | XP_016884319.1 | 41 | 4812 | |||
GenBank mRNA | AB210002.1 | - | 55 | 6626 | ||
AF012872.1 | - | 54 | 6395 | |||
AK094594.1 | - | 1 | 2040 | |||
AK095811.1 | - | 13 | 3026 | |||
AK124692.1 | - | 10 | 3831 | |||
AK302424.1 | - | 55 | 6584 | |||
BC018120.1 | - | 25 | 2973 | |||
BC053654.1 | - | 24 | 2949 | |||
L36151.1 | - | 23 | 2942 |