Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oprd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013622.3 | NP_038650.3 | 2 | 480 | ||
GenBank mRNA | BC137958.1 | - | 2 | 407 | 8860977 | |
BC137969.1 | - | 2 | 407 | 8860988 | ||
L06322.1 | - | 2 | 437 | |||
L07271.1 | - | 2 | 407 | |||
L11064.1 | - | 2 | 390 | |||
S65335.1 | - | 2 | 406 | |||
S66181.1 | - | 2 | 358 |
Set Membership: |
G-protein coupled receptor |