Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ID3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002167.4 | NP_002158.3 | 1 | 600 | ||
GenBank mRNA | A17546.1 | - | 1 | 355 | ||
A17548.1 | - | 1 | 232 | |||
AK225965.1 | - | 1 | 335 | |||
AK290003.1 | - | 1 | 337 | |||
BC003107.1 | - | 1 | 318 | 3543936 | ||
BM921819.1 | - | 631 | ||||
BT006791.1 | - | 1 | 232 | |||
X66924.1 | - | 1 | 355 | |||
X69111.1 | - | 1 | 599 |