Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc22a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008766.3 | NP_032792.2 | 3 | 901 | ||
GenBank mRNA | AK035971.1 | - | 3 | 719 | ||
BC021647.1 | - | 3 | 869 | 4163278 | ||
U52842.1 | - | 3 | 782 |