Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ID4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001546.3 | NP_001537.1 | 3 | 1800 | ||
GenBank mRNA | AJ420553.1 | - | 1 | 54 | ||
AK130851.1 | - | 1 | 173 | |||
BC014941.1 | - | 3 | 1784 | 4552357 |