Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sqstm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290769.1 | NP_001277698.1 | 3 | 480 | ||
NM_011018.3 | NP_035148.1 | 3 | 480 | |||
GenBank mRNA | BC006019.1 | - | 3 | 477 | 3487289 | |
E08146.1 | - | 3 | 438 | |||
U40930.1 | - | 3 | 450 | |||
U57413.1 | - | 3 | 442 |