Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cldn11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008770.3 | NP_032796.1 | 3 | 794 | ||
GenBank mRNA | AF124426.1 | - | 3 | 564 | ||
AK005088.1 | - | 3 | 769 | |||
AK005171.1 | - | 3 | 769 | |||
AK161698.1 | - | 3 | 769 | |||
BC021659.1 | - | 3 | 746 | 4190031 | ||
U19582.1 | - | 3 | 723 |