Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cldn11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008770.3 | NP_032796.1 | 3 | 1723 | ||
GenBank mRNA | AK005088.1 | - | 3 | 1698 | ||
AK005171.1 | - | 3 | 1696 | |||
AK161698.1 | - | 3 | 1699 | |||
BB798247.1 | - | 1 | 248 | |||
BC021659.1 | - | 3 | 1677 | 4190031 | ||
U19582.1 | - | 3 | 1651 |