Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Otx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286481.1 | NP_001273410.1 | 5 | 1202 | ||
NM_001286482.1 | NP_001273411.1 | 5 | 1178 | |||
NM_001286483.1 | NP_001273412.1 | 4 | 1018 | |||
NM_144841.4 | NP_659090.1 | 3 | 1100 | |||
GenBank mRNA | AK053741.1 | - | 4 | 1015 | ||
AK081535.1 | - | 4 | 1019 | |||
AK087527.1 | - | 4 | 1016 | |||
BC017609.1 | - | 3 | 1100 | 4527414 | ||
BC027104.1 | - | 5 | 1060 | 5359966 | ||
BC029667.1 | - | 5 | 1064 | 5362013 |