Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2rx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008771.3 | NP_032797.3 | 1 | 237 | ||
GenBank mRNA | AK035304.1 | - | 1 | 238 | ||
BC015084.1 | - | 1 | 251 | 4189541 | ||
X84896.1 | - | 1 | 231 |