Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2rx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011026.2 | NP_035156.2 | 6 | 662 | ||
GenBank mRNA | AF089751.1 | - | 6 | 596 | ||
AJ251459.1 | - | 6 | 576 | |||
AJ251461.1 | - | 6 | 576 | |||
AK150351.1 | - | 6 | 652 | |||
AK162820.1 | - | 6 | 628 | |||
AK163104.1 | - | 6 | 650 | |||
BC002099.1 | - | 6 | 636 | |||
JX417451.1 | - | 6 | 622 | |||
JX674050.1 | - | 2 | 230 |