Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2rx6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159561.1 | NP_001153033.1 | 2 | 358 | ||
NM_011028.2 | NP_035158.2 | 2 | 358 | |||
GenBank mRNA | AB010883.1 | - | 2 | 243 | ||
AK054195.1 | - | 311 | ||||
AK081308.1 | - | 2 | 234 | |||
AK138541.1 | - | 2 | 210 | |||
BC117713.2 | - | 2 | 220 | 40088045 |