Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pak3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195046.1 | NP_001181975.1 | 6, 7 | 597 | ||
NM_001195047.1 | NP_001181976.1 | 5, 6 | 552 | |||
NM_001195048.1 | NP_001181977.1 | 6, 7 | 707 | |||
NM_001195049.1 | NP_001181978.1 | 8, 9 | 971 | |||
NM_008778.3 | NP_032804.2 | 5, 6 | 580 | |||
GenBank mRNA | AF082297.1 | - | 4, 5 | 494 | ||
AJ496262.1 | - | 4, 5 | 462 | |||
AJ496263.1 | - | 5, 6 | 507 | |||
AK031853.1 | - | 6, 7 | 588 | |||
AK158893.1 | - | 6, 7 | 707 | |||
AM943853.1 | - | 5, 6 | 525 | |||
AM943854.1 | - | 6, 7 | 570 | |||
BC053403.1 | - | 5, 6 | 587 | 30060082 | ||
U39738.1 | - | 8, 10 | 978 |