Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pappa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021362.1 | NP_067337.1 | 2, 3 | 1845 | ||
GenBank mRNA | AF258461.1 | - | 1, 2 | 706 | ||
AF439513.1 | - | 2, 3 | 1240 | |||
AF439514.1 | - | 3, 4 | 1327 |