Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IDI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317955.1 | NP_001304884.1 | 6 | 1127 | ||
NM_001317956.1 | NP_001304885.1 | 5 | 1261 | |||
NM_001317957.1 | NP_001304886.1 | 4 | 1088 | |||
NM_004508.3 | NP_004499.2 | 5 | 1144 | |||
NR_134300.1 | 4 | 876 | ||||
NR_134301.1 | 4 | 971 | ||||
XM_011519457.1 | XP_011517759.1 | 5 | 1991 | |||
XM_017016191.1 | XP_016871680.1 | 4 | 1818 | |||
GenBank mRNA | AF271720.1 | - | 4 | 629 | ||
AK222875.1 | - | 5 | 664 | |||
AK303669.1 | - | 5 | 898 | |||
AK311110.1 | - | 4 | 520 | |||
BC005247.2 | - | 4 | 642 | |||
BC006999.2 | - | 4 | 642 | |||
BC019227.2 | - | 4 | 642 | |||
BC022418.2 | - | 4 | 642 | |||
BC025375.2 | - | 4 | 642 | |||
BC057827.1 | - | 6 | 1115 | 30337530 | ||
BC107893.1 | - | 5 | 668 | |||
BT006761.1 | - | 4 | 613 | |||
BX537663.1 | - | 4 | 3688 | |||
BX648472.1 | - | 4 | 1607 | |||
X17025.1 | - | 5 | 663 |