Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pax5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008782.2 | NP_032808.1 | 3 | 315 | ||
GenBank mRNA | JX065130.1 | - | 2 | 196 | ||
M97013.1 | - | 3 | 315 |