Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001103165.1 | NP_001096635.1 | 11 | 998 | ||
NM_001103166.1 | NP_001096636.1 | 10 | 905 | |||
NM_001174073.1 | NP_001167544.1 | 10 | 893 | |||
NM_011042.2 | NP_035172.2 | 11 | 998 | |||
GenBank mRNA | AK012297.1 | - | 11 | 845 | ||
AK161527.1 | - | 10 | 741 | |||
AK169268.1 | - | 11 | 1092 | |||
AK213763.1 | - | 3 | 126 | |||
BC020478.1 | - | 9 | 650 | |||
EU604548.1 | - | 10 | 753 | |||
L19661.1 | - | 11 | 885 |