Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcsk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013628.2 | NP_038656.1 | 3, 4 | 468 | ||
GenBank mRNA | AK043742.1 | - | 3, 4 | 540 | ||
BC108982.1 | - | 3, 4 | 446 | 40047652 | ||
BC108983.1 | - | 3, 4 | 446 | 40047655 | ||
M58589.1 | - | 3, 4 | 468 | |||
M69196.1 | - | 3, 4 | 574 |